Benzyl propyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%

Supplier: Ambeed
Warning

A683082-1G A683082-250MG A683082-5G
77198-674EA 98 USD
77198-674 77198-676 77198-678
Benzyl propyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 97%
Benzyl propyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₃H₃₀BNO₄
MW: 395.29 g/mol
MDL Number: MFCD21609688
CAS Number: 2096332-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR