Methyl 4-amino-3-bromo-5-chlorobenzoate 97%

Supplier: Ambeed
Warning

A682975-100MG A682975-1G A682975-250MG
77198-668EA 10 USD
77198-668 77198-670 77198-672
Methyl 4-amino-3-bromo-5-chlorobenzoate 97%
Methyl 4-amino-3-bromo-5-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MDL Number: MFCD08436084
CAS Number: 914636-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR