3-(2-Thienyl)propanoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 3-(2-Thienyl)propionic Acid

A682739-10G A682739-1G A682739-25G A682739-5G
77198-660EA 144.2 USD
77198-660 77198-662 77198-664 77198-666
3-(2-Thienyl)propanoic acid 97%
3-(2-Thienyl)propanoic acid

Precaución: for Research Use Only

Formula: C₇H₈O₂S
MW: 156.199 g/mol
MDL Number: MFCD00047093
CAS Number: 5928-51-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR