CAY-10441 97%

Supplier: Ambeed
Warning

Synonyms: RO-1138452, N-(4-(4-Isopropoxybenzyl)phenyl)-4,5-dihydro-1H-imidazol-2-amine, RO1138452, 4,5-dihydro-N-[4-[[4-(1-methylethoxy)phenyl]methyl]phenyl]-1H-imadazol-2-amine

A682702-100MG A682702-10MG A682702-1MG A682702-25MG A682702-50MG A682702-5MG
77198-648EA 313 USD
77198-648 77198-650 77198-652 77198-654 77198-656 77198-658
CAY-10441 97%
CAY-10441

Precaución: for Research Use Only

Formula: C₁₉H₂₃N₃O
CAS Number: 221529-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR