(S)-2-Amino-1-(pyrrolidin-1-yl)propane-1-thione hydrochloride 97%

Supplier: Ambeed
Warning

A681426-100MG A681426-1G A681426-250MG
77198-638EA 43 USD
77198-638 77198-640 77198-642
(S)-2-Amino-1-(pyrrolidin-1-yl)propane-1-thione hydrochloride 97%
(S)-2-Amino-1-(pyrrolidin-1-yl)propane-1-thione hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅ClN₂S
MDL Number: MFCD09908178
CAS Number: 184360-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR