(2R,3R,4S)-4-(Benzo[d][1,3]dioxol-5-yl)-1-(2-(dibutylamino)-2-oxoethyl)-2-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Atrasentan HCl
A679665-1MG
A679665-2MG
77170-578EA
79.31
USD
77170-578
77170-580
(2R,3R,4S)-4-(Benzo[d][1,3]dioxol-5-yl)-1-(2-(dibutylamino)-2-oxoethyl)-2-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid hydrochloride 98%
(2R,3R,4S)-4-(Benzo[d][1,3]dioxol-5-yl)-1-(2-(dibutylamino)-2-oxoethyl)-2-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₂₉H₃₉ClN₂O₆ MW: 547.08 g/mol |
MDL Number:
MFCD13194884 CAS Number: 195733-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...