1,2,4,5-Tetrafluorobenzene 98%

Supplier: Ambeed
Danger

A679444-1G A679444-25G A679444-5G
77198-628EA 5.15 USD
77198-628 77198-630 77198-632
1,2,4,5-Tetrafluorobenzene 98%
1,2,4,5-Tetrafluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₄
MW: 150.08 g/mol
MDL Number: MFCD00000307
CAS Number: 327-54-8
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR