5-Bromo-2-fluoro-3-iodopyridine 97%

Supplier: Ambeed
Warning

A678470-100MG A678470-1G A678470-250MG
77198-622EA 10 USD
77198-622 77198-624 77198-626
5-Bromo-2-fluoro-3-iodopyridine 97%
5-Bromo-2-fluoro-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFIN
MDL Number: MFCD13185373
CAS Number: 1214376-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR