1,3-Dimethyl-2-oxo-2,3-dihydro-1H-benzimidazole-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A677184-100MG A677184-1G
77198-618EA 30 USD
77198-618 77198-620
1,3-Dimethyl-2-oxo-2,3-dihydro-1H-benzimidazole-5-carbaldehyde 95%
1,3-Dimethyl-2-oxo-2,3-dihydro-1H-benzimidazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD00772536
CAS Number: 55241-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR