2-Amino-6-methylnicotinamide 97%

Supplier: Ambeed
Warning

A676827-100MG A676827-1G A676827-5G
77198-612EA 141.11 USD
77198-612 77198-614 77198-616
2-Amino-6-methylnicotinamide 97%
2-Amino-6-methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.16 g/mol
MDL Number: MFCD11107426
CAS Number: 100524-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR