5-Fluoro-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A676146-100MG A676146-1G A676146-250MG
77198-606EA 333.72 USD
77198-606 77198-608 77198-610
5-Fluoro-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde 95%
5-Fluoro-1H-pyrrolo[3,2-b]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MW: 164.13 g/mol
MDL Number: MFCD12962581
CAS Number: 1190322-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR