tert-Butyl 2-(hydrazinecarbonyl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A675727-1G A675727-5G
77170-572EA 11.33 USD
77170-572 77170-574
tert-Butyl 2-(hydrazinecarbonyl)pyrrolidine-1-carboxylate 97%
tert-Butyl 2-(hydrazinecarbonyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉N₃O₃
MW: 229.27 g/mol
MDL Number: MFCD06738833
CAS Number: 359803-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR