5-Fluorobenzofuran-2(3H)-one 97%

Supplier: Ambeed
Warning

A675154-100MG A675154-250MG
77198-602EA 47 USD
77198-602 77198-604
5-Fluorobenzofuran-2(3H)-one 97%
5-Fluorobenzofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₈H₅FO₂
MDL Number: MFCD28163523
CAS Number: 886985-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR