4-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%

Supplier: Ambeed
Warning

A674958-1G A674958-5G
77170-568EA 31 USD
77170-568 77170-570
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%
4-Bromo-5'-phenyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₄H₁₇Br
MDL Number: MFCD26407506
CAS Number: 116941-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR