But-3-en-1-ylboronic acid 97% + stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A673302-1G A673302-250MG A673302-5G
77170-544EA 62 USD
77170-544 77170-546 77170-548
But-3-en-1-ylboronic acid 97% + stabilised
But-3-en-1-ylboronic acid

Precaución: for Research Use Only

Formula: C₄H₉BO₂
MDL Number: MFCD09836235
CAS Number: 379669-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR