Methyl 3,5-dichloro-2-methoxybenzoate 95%

Supplier: Ambeed
Warning

A672650-100MG A672650-1G A672650-250MG A672650-5G
77198-594EA 24 USD
77198-594 77198-596 77198-598 77198-600
Methyl 3,5-dichloro-2-methoxybenzoate 95%
Methyl 3,5-dichloro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
MDL Number: MFCD14704683
CAS Number: 64122-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR