1,2-Dibromo-3,5-difluorobenzene 98%

Supplier: Ambeed
Warning

A670554-1G A670554-5G A670554-25G
77198-588EA 5.15 USD
77198-588 77198-590 77747-359
1,2-Dibromo-3,5-difluorobenzene 98%
1,2-Dibromo-3,5-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MW: 271.89 g/mol
MDL Number: MFCD00042559
CAS Number: 139215-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR