Diethyl-1,4-phenylenediacetate 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2,2-(1,4-phenylene)diacetate, 1,4-Phenylenediacetic Acid Diethyl Ester

A670491-1G A670491-5G
77198-584EA 29 USD
77198-584 77198-586
Diethyl-1,4-phenylenediacetate 98%
Diethyl-1,4-phenylenediacetate

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
MDL Number: MFCD00032366
CAS Number: 36076-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR