4-(2-Fluorophenyl)tetrahydro-2H-pyran-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A669725-100MG A669725-1G A669725-250MG
77198-576EA 79.31 USD
77198-576 77198-578 77198-580
4-(2-Fluorophenyl)tetrahydro-2H-pyran-4-amine hydrochloride 97%
4-(2-Fluorophenyl)tetrahydro-2H-pyran-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClFNO
MW: 231.69 g/mol
MDL Number: MFCD22375732
CAS Number: 1380300-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR