1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea 96%

Supplier: Ambeed
Warning

A669183-100G A669183-1G A669183-25G A669183-500G A669183-5G
77198-564EA 217.33 USD
77198-564 77198-566 77198-568 77198-570 77198-572
1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea 96%
1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₄S
MW: 206.22 g/mol
MDL Number: MFCD00674104
CAS Number: 34840-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR