4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 98%

Supplier: Ambeed
Warning

A668035-100MG A668035-1G A668035-250MG
77198-552EA 31 USD
77198-552 77198-554 77198-556
4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile 98%
4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MW: 244.09 g/mol
MDL Number: MFCD18719940
CAS Number: 2009346-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR