1,3-Diphenyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A666832-25G A666832-5G
77198-548EA 512.94 USD
77198-548 77198-550
1,3-Diphenyl-1H-pyrazole-4-carbaldehyde 97%
1,3-Diphenyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O
MW: 248.29 g/mol
MDL Number: MFCD00525714
CAS Number: 21487-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR