(R)-tert-Butyl (1-(2-bromo-4-chlorophenyl)ethyl)carbamate 95%

Supplier: Ambeed
Warning

A666136-1G A666136-250MG
77170-514EA 56.65 USD
77170-514 77170-516
(R)-tert-Butyl (1-(2-bromo-4-chlorophenyl)ethyl)carbamate 95%
(R)-tert-Butyl (1-(2-bromo-4-chlorophenyl)ethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrClNO₂
MW: 334.64 g/mol
MDL Number: MFCD28805510
CAS Number: 1799438-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR