4-Bromo-6-(tert-butyl)dibenzo[b,d]furan 95%
Supplier: Ambeed
|
Warning
|
A666043-1G
A666043-5G
77170-510EA
28
USD
77170-510
77170-512
4-Bromo-6-(tert-butyl)dibenzo[b,d]furan 95%
4-Bromo-6-(tert-butyl)dibenzo[b,d]furan
Precaución: for Research Use Only
Formula:
C₁₆H₁₅BrO MW: 303.19 g/mol |
MDL Number:
MFCD29919031 CAS Number: 1438391-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...