8-Bromo-1-tetralone 96%

Supplier: Ambeed
Warning

Synonyms: 8-Bromo-3,4-dihydronaphthalen-1(2H)-one

A665637-100MG A665637-1G A665637-250MG
77170-498EA 38.11 USD
77170-498 77170-500 77170-502
8-Bromo-1-tetralone 96%
8-Bromo-1-tetralone

Precaución: for Research Use Only

Formula: C₁₀H₉BrO
MW: 225.08 g/mol
MDL Number: MFCD06796575
CAS Number: 651735-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR