Methyl 2-(bromomethyl)-4-fluorobenzoate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl α-bromo-4-fluoro-o-toluate, 2-(Methoxycarbonyl)-5-fluorobenzyl bromide

A663951-100MG A663951-250MG
77170-486EA 7 USD
77170-486 77170-488
Methyl 2-(bromomethyl)-4-fluorobenzoate 97%
Methyl 2-(bromomethyl)-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
MDL Number: MFCD11505951
CAS Number: 157652-28-3
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR