1-(3-Chlorophenyl)cyclopropanamine 95%

Supplier: Ambeed
Warning

A661685-1G A661685-250MG
77170-476EA 74.16 USD
77170-476 77170-478
1-(3-Chlorophenyl)cyclopropanamine 95%
1-(3-Chlorophenyl)cyclopropanamine

Precaución: for Research Use Only

Formula: C₉H₁₀ClN
MW: 167.64 g/mol
MDL Number: MFCD07374477
CAS Number: 474709-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR