Methacholine chloride 98%

Supplier: Ambeed
Warning

Synonyms: Mecholyl Chloride, O-Acetyl-beta-methylcholine Chloride, Acetyl-β-methylcholine chloride

A661665-100MG A661665-25MG
77198-524EA 6.18 USD
77198-524 77198-526
Methacholine chloride 98%
Methacholine chloride

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO₂
MW: 195.69 g/mol
MDL Number: MFCD00011817
CAS Number: 62-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR