Ethyl 4-((4-methoxybenzyl)amino)-2-(methylthio)pyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A659647-1G A659647-250MG
77170-458EA 35.02 USD
77170-458 77170-460
Ethyl 4-((4-methoxybenzyl)amino)-2-(methylthio)pyrimidine-5-carboxylate 95%
Ethyl 4-((4-methoxybenzyl)amino)-2-(methylthio)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₉N₃O₃S
MW: 333.40 g/mol
MDL Number: MFCD05156568
CAS Number: 211230-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR