5-Bromo-3,3-dimethylindolin-2-one 98%

Supplier: Ambeed
Warning

A659376-1G A659376-250MG A659376-5G
77170-450EA 223.51 USD
77170-450 77170-452 77170-454
5-Bromo-3,3-dimethylindolin-2-one 98%
5-Bromo-3,3-dimethylindolin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD11919699
CAS Number: 120902-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR