2-Chloro-3-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A659178-10G A659178-250MG A659178-25G A659178-5G A659178-1G
77198-496EA 114.33 USD
77198-496 77198-498 77198-500 77198-502 77747-345
2-Chloro-3-(trifluoromethyl)benzaldehyde 95%
2-Chloro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
MDL Number: MFCD00792414
CAS Number: 93118-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR