3-Bromo-4,5-dichlorophenol 97%

Supplier: Ambeed
Warning

A658694-100MG A658694-1G A658694-250MG
77198-490EA 46 USD
77198-490 77198-492 77198-494
3-Bromo-4,5-dichlorophenol 97%
3-Bromo-4,5-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MDL Number: MFCD28737852
CAS Number: 1805523-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR