5-Acetyl-2-amino-4-methylpyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Amino-4-methylpyrimidin-5-yl)ethanone

A658115-1G A658115-250MG
77198-476EA 62.83 USD
77198-476 77198-478
5-Acetyl-2-amino-4-methylpyrimidine 97%
5-Acetyl-2-amino-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MW: 151.17 g/mol
MDL Number: MFCD00052621
CAS Number: 66373-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR