2-Bromo-4-ethoxy-1-methylbenzene 97%

Supplier: Ambeed
Warning

A658074-100G A658074-25G A658074-5G
77198-470EA 86.52 USD
77198-470 77198-472 77198-474
2-Bromo-4-ethoxy-1-methylbenzene 97%
2-Bromo-4-ethoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.08 g/mol
MDL Number: MFCD28142603
CAS Number: 1445601-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR