1,10-Bis(acryloyloxy)decane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A657386-100G A657386-25G A657386-5G
77198-464EA 110.21 USD
77198-464 77198-466 77198-468
1,10-Bis(acryloyloxy)decane 98% stabilized
1,10-Bis(acryloyloxy)decane

Precaución: for Research Use Only

Formula: C₁₆H₂₆O₄
MW: 282.38 g/mol
MDL Number: MFCD00080453
CAS Number: 13048-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR