2-Chloro-4-methoxy-5-methylpyrimidine 95%

Supplier: Ambeed
Warning

A657370-100MG A657370-250MG
77170-444EA 44.29 USD
77170-444 77170-446
2-Chloro-4-methoxy-5-methylpyrimidine 95%
2-Chloro-4-methoxy-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MDL Number: MFCD20483333
CAS Number: 135292-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR