2-Morpholino-1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone 98+%

Supplier: Ambeed
Warning

A656801-1G A656801-250MG A656801-5G
77170-438EA 193.64 USD
77170-438 77170-440 77170-442
2-Morpholino-1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone 98+%
2-Morpholino-1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₂₀H₂₉BN₂O₄
MW: 372.27 g/mol
MDL Number: MFCD28384365
CAS Number: 1704067-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR