1,3-Di-o-tolylguanidine 98%

Supplier: Ambeed
Warning

A656782-100G A656782-25G A656782-500G
77198-458EA 32.96 USD
77198-458 77198-460 77198-462
1,3-Di-o-tolylguanidine 98%
1,3-Di-o-tolylguanidine

Precaución: for Research Use Only

Formula: C₁₅H₁₇N₃
MW: 239.32 g/mol
MDL Number: MFCD00008513
CAS Number: 97-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR