5,6-Dihydroxyisobenzofuran-1(3H)-one 97%

Supplier: Ambeed
Warning

A654733-100MG A654733-250MG A654733-50MG
77198-444EA 92 USD
77198-444 77198-446 77198-448
5,6-Dihydroxyisobenzofuran-1(3H)-one 97%
5,6-Dihydroxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₆O₄
MW: 166.13 g/mol
MDL Number: MFCD15146112
CAS Number: 53766-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR