(S)-2-Amino-4-(methylamino)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

A654723-5G A654723-250MG
77170-436EA 188 USD
77170-436 77893-313
(S)-2-Amino-4-(methylamino)-4-oxobutanoic acid 98%
(S)-2-Amino-4-(methylamino)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₃
MDL Number: MFCD16251112
CAS Number: 7175-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR