2,3,6-Trichloro-5-(chloromethyl)pyridine 95%

Supplier: Ambeed
Danger

A653693-100MG A653693-1G A653693-250MG
77170-422EA 133.9 USD
77170-422 77170-424 77170-426
2,3,6-Trichloro-5-(chloromethyl)pyridine 95%
2,3,6-Trichloro-5-(chloromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₄N
MW: 230.90 g/mol
CAS Number: 52465-59-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR