1-Phenylcyclopropanamine 96%

Supplier: Ambeed
Danger

Synonyms: 1-Phenylcyclopropylamine

A653334-10G A653334-1G A653334-5G
77170-416EA 461.44 USD
77170-416 77170-418 77170-420
1-Phenylcyclopropanamine 96%
1-Phenylcyclopropanamine

Precaución: for Research Use Only

Formula: C₉H₁₁N
MW: 133.19 g/mol
MDL Number: MFCD05863520
CAS Number: 41049-53-0
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR