(E)-Ethyl cinnamate 97%

Supplier: Ambeed
Warning

A651672-100G A651672-25G
77198-432EA 23 USD
77198-432 77198-434
(E)-Ethyl cinnamate 97%
(E)-Ethyl cinnamate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MDL Number: MFCD00009189
CAS Number: 4192-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR