3-Acetyl-1-methylpyrrolidin-2-one 97%

Supplier: Ambeed
Warning

A648158-100MG A648158-250MG
77198-424EA 89.61 USD
77198-424 77198-426
3-Acetyl-1-methylpyrrolidin-2-one 97%
3-Acetyl-1-methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
MDL Number: MFCD02683822
CAS Number: 60044-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR