2-Iodoisonicotinic acid 95%

Supplier: Ambeed
Warning

A648117-1G A648117-250MG A648117-5G
77170-396EA 28 USD
77170-396 77170-398 77893-292
2-Iodoisonicotinic acid 95%
2-Iodoisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄INO₂
MDL Number: MFCD06800803
CAS Number: 58481-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR