6-Chloro-2-(methylthio)benzo[d]thiazole 95%

Supplier: Ambeed
Warning

A647237-1G A647237-250MG A647237-5G
77198-416EA 127.72 USD
77198-416 77198-418 77198-420
6-Chloro-2-(methylthio)benzo[d]thiazole 95%
6-Chloro-2-(methylthio)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS₂
MW: 215.72 g/mol
CAS Number: 3507-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR