2-Chloroisonicotinic acid hydrazide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-pyridinecarbohydrazide

A646046-100MG A646046-1G A646046-1MG A646046-5G
77170-380EA 16 USD
77170-380 77170-382 77170-384 77170-386
2-Chloroisonicotinic acid hydrazide 98%
2-Chloroisonicotinic acid hydrazide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MDL Number: MFCD00221428
CAS Number: 58481-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR