1-Benzyl-5-oxopyrrolidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-5-oxo-3-pyrrolidinecarboxylic Acid

A645534-1G A645534-25G
77198-408EA 31.93 USD
77198-408 77198-410
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid 98%
1-Benzyl-5-oxopyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD00085749
CAS Number: 5733-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR