(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid 95%

Supplier: Ambeed
Warning

A644676-100MG A644676-250MG A644676-1G
77170-372EA 5.15 USD
77170-372 77170-374 77684-524
(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid 95%
(2R,4S)-5-([1,1'-Biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpentanoic acid

Precaución: For research use only.

Formula: C₂₃H₂₉NO₄
MW: 383.48 g/mol
MDL Number: MFCD27955981
CAS Number: 1012341-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR