Dimethyl(octadecyl)silane 95+%

Supplier: Ambeed
Warning

A644283-1G A644283-250MG A644283-5G
77170-366EA 32 USD
77170-366 77170-368 77170-370
Dimethyl(octadecyl)silane 95+%
Dimethyl(octadecyl)silane

Precaución: for Research Use Only

Formula: C₂₀H₄₄Si
MW: 312.64 g/mol
MDL Number: MFCD00010224
CAS Number: 32395-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR